• USA Home
  • Search Results

  • tyrala2522773008511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tyr-Ala

1 Products
Image of Tyr-Ala
CAS Number:
Empirical Formula (Hill Notation):
 C12H16N2O4
Molecular Weight:
 252.27
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
T5129 Tyr-Ala,