• USA Home
  • Search Results

  • tyramine137185167211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tyramine

3 Products
Synonym:   2-(4-Hydroxyphenyl)ethylamine, 4-(2-Aminoethyl)phenol, 4-Hydroxyphenethylamine
Image of Tyramine
CAS Number:
Linear Formula:
 HOC6H4CH2CH2NH2
Molecular Weight:
 137.18
 Beilstein/REAXYS Number:
 1099914
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Titertest
  Special Grade
Show More