• USA Home
  • Search Results

  • u69593356509674475111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

U-69593

1 Products
Synonym:   (+)-(5α,7α,8β)-N-Methyl-N-[7-(1-pyrrolidinyl)-1-oxaspiro[4.5]dec-8-yl]-benzeneacetamide, U69593
Image of U-69593
CAS Number:
Empirical Formula (Hill Notation):
 C22H32N2O2
Molecular Weight:
 356.50
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
Show More

Product #
Description
U103 U-69593, solid,