• USA Home
  • Search Results

  • umbelliferone162149335611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Umbelliferone

4 Products
Synonym:   7-Hydroxy-2H-1-benzopyran-2-one, 7-Hydroxycoumarin
Image of Umbelliferone
CAS Number:
Empirical Formula (Hill Notation):
 C9H6O3
Molecular Weight:
 162.14
 Beilstein/REAXYS Number:
 127683
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Application(s)
  Product Line
  Solubility
Show More