• USA Home
  • Search Results

  • uniconazole291788365722111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Uniconazole

2 Products
Synonym:   (E)-(RS)-1-(4-Chlorophenyl)-4,4-dimethyl-2-(1H-1,2,4-triazol-1-yl)pent-1-en-3-ol
Image of Uniconazole
CAS Number:
Empirical Formula (Hill Notation):
 C15H18ClN3O
Molecular Weight:
 291.78
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Anal. Standard Form
  Shelf Life
  Physical Form
  Special Grade
  Chemical Composition
Show More