• USA Home
  • Search Results

  • uracil112096622811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Uracil

4 Products
Synonym:   2,4(1H,3H)-Pyrimidinedione, 2,4-Dihydroxypyrimidine, 2,4-Pyrimidinediol
Image of Uracil
CAS Number:
Empirical Formula (Hill Notation):
 C4H4N2O2
Molecular Weight:
 112.09
 Beilstein/REAXYS Number:
 507828
 EC Number:
 MDL number:
 NACRES:
 NA.51
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Product Line
  Application(s)
  Assay
Show More