• USA Home
  • Search Results

  • urea13c15n263035806983311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Urea-13C,15N2

1 Products
Image of Urea-<SUP>13</SUP>C,<SUP>15</SUP>N<SUB>2</SUB>
CAS Number:
Linear Formula:
 H215N13CO15NH2
Molecular Weight:
 63.03
 MDL number:
 NACRES:
 NA.12
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Isotopic Purity
  Assay
Show More