• USA Home
  • Search Results

  • urea15n26204206780311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Urea-15N2

5 Products
Synonym:   15N Labeled Urea, 15N Labeled urea, Carbamide-15N2, Carbonyldiamide-15N2
Image of Urea-<SUP>15</SUP>N<SUB>2</SUB>
CAS Number:
Linear Formula:
 H215NCO15NH2
Molecular Weight:
 62.04
 MDL number:
 NACRES:
 NA.12
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Molecular weight
  Isotopic Purity
Show More