• USA Home
  • Search Results

  • urea60065713611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Urea

25 Products
Synonym:   Carbamide, Carbonyl diamide, Diaminomethanone, Carbonyldiamine, Urea
Image of Urea
CAS Number:
Linear Formula:
 CO(NH2)2
Molecular Weight:
 60.06
 EC Index Number
 200-315-5
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  UV-Absorption Values
  Quality clarifier
  Restriction
  Package quantity
  Loss
  Titertest
  Purity Grade
  Product Line
  Anal. Standard Form
  Suitability
  Absorption
  Vapor pressure
  Density
  Bulk Density
  pH Value
  Melting Point
  Potency
  Physical Form
  Solubility
  Assay
Show More

Product #
Description
U5378 Urea, powder, BioReagent, for molecular biology, suitable for cell culture,
U0631 Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O),
U6504 Urea, for electrophoresis,
U5128 Urea, ACS reagent, 99.0-100.5%,
U4883 Urea, 8 M (after reconstitution with 16 mL high purity water),
U4884 Urea, meets USP testing specifications,
U1250 Urea, ReagentPlus®, ≥99.5%, pellets,
15604 Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance),
51456 Urea, BioUltra, for molecular biology, ≥99% (T),
33247 Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%,
1.37030 Urea, cryst. EMPROVE® EXPERT Ph Eur,BP,JP,USP,ACS,
1.08486 Urea, cryst. EMPROVE® ESSENTIAL Ph Eur,BP,USP,JP,
1.08484 Urea, beads,
1.08487 Urea, GR for analysis ACS,Reag. Ph Eur,
1.08488 Urea, for biochemistry,
1706698 Urea, United States Pharmacopeia (USP) Reference Standard,
8.18710 Urea, for synthesis,
PHR1406 Urea, Pharmaceutical Secondary Standard; Certified Reference Material,
56180 Urea, analytical standard,
U0600000 Urea, European Pharmacopoeia (EP) Reference Standard,
NIST2141 Urea, NIST® SRM® 2141,
NIST2152 Urea, NIST® SRM® 2152, combustion calorimetric standard,
NIST912A Urea, NIST® SRM® 912a, clinical standard,
BP350 Urea, British Pharmacopoeia (BP) Reference Standard,
RDD034 Urea, free-flowing, Redi-Dri, Bioreagent, ≥98%, New