• USA Home
  • Search Results

  • uricacid168116993211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Uric acid

3 Products
Synonym:   2,6,8-Trihydroxypurine
Image of Uric acid
CAS Number:
Empirical Formula (Hill Notation):
 C5H4N4O3
Molecular Weight:
 168.11
 Beilstein/REAXYS Number:
 156158
 EC Number:
 MDL number:
 NACRES:
 NA.55
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
Show More