• USA Home
  • Search Results

  • uridine244205896811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Uridine

5 Products
Synonym:   1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside
Image of Uridine
CAS Number:
Empirical Formula (Hill Notation):
 C9H12N2O6
Molecular Weight:
 244.20
 Beilstein/REAXYS Number:
 754902
 EC Number:
 MDL number:
 NACRES:
 NA.51
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Product Line
  Biological Source
  Solubility
  Special Grade
Show More