• USA Home
  • Search Results

  • urokinasefromhumankidneycells12345903953611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Urokinase from human kidney cells

1 Products
Synonym:   u-Plasminogen activator
CAS Number:
 EC Number:
 Enzyme Commission (EC) Number:
 3.4.21.73  ( BRENDA | IUBMB )
 MDL number:
 eCl@ss:
 32160410
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More