• USA Home
  • Search Results

  • ursodeoxycholicacid3925712813211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ursodeoxycholic acid

4 Products
Synonym:   3α,7β-Dihydroxy-5β-cholan-24-oic acid, 5β-Cholan-24-oic acid-3α,7β-diol, 7β-Hydroxylithocholic acid, UDCS
Image of Ursodeoxycholic acid
CAS Number:
Empirical Formula (Hill Notation):
 C24H40O4
Molecular Weight:
 392.57
 Beilstein/REAXYS Number:
 3219888
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Anal. Standard Form
Show More