• USA Home
  • Search Results

  • valericacid1021310952411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Valeric acid

5 Products
Synonym:   n-Valeric acid, Pentanoic acid
Image of Valeric acid
CAS Number:
Linear Formula:
 CH3(CH2)3COOH
Molecular Weight:
 102.13
 Beilstein/REAXYS Number:
 969454
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Document Links
  Shelf Life
  Allergen
  Organoleptic property
  Application(s)
  Biological Source
  Assay
  Agency/Method
Show More