• USA Home
  • Search Results

  • valifenalate3988828315990011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Valifenalate

1 Products
Synonym:   N-[(1-Methylethoxy)carbonyl]-L-valyl-3-(4-chlorophenyl)-β-alanine methyl ester, Methyl N-(isopropoxycarbonyl)-L-valyl-(3RS)-3-(4-chlorophenyl)-β-alaninate
Image of Valifenalate
CAS Number:
Empirical Formula (Hill Notation):
 C19H27ClN2O5
Molecular Weight:
 398.88
 Beilstein/REAXYS Number:
 11327531
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More