• USA Home
  • Search Results

  • vancomycinhydrochloridefromstreptomycesorientalis148571140493911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Vancomycin hydrochloride from Streptomyces orientalis

4 Products
Image of Vancomycin hydrochloride from <I>Streptomyces orientalis</I>
CAS Number:
Empirical Formula (Hill Notation):
 C66H75Cl2N9O24 · HCl
Molecular Weight:
 1485.71
 Beilstein/REAXYS Number:
 3704657
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Biological Source
  Color
  Mode of Action
  Activity Spectrum
  Physical Form
  Solubility
  Potency
Show More