• USA Home
  • Search Results

  • vanillin1521512133511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Vanillin

11 Products
Synonym:   4-Hydroxy-3-methoxybenzaldehyde, Vanillic aldehyde
Image of Vanillin
CAS Number:
Linear Formula:
 4-(HO)C6H3-3-(OCH3)CHO
Molecular Weight:
 152.15
 Beilstein/REAXYS Number:
 472792
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Shelf Life
  Special Grade
  Allergen
  Organoleptic property
  pH Value
  Vapor pressure
  Transition temperature
  Product Line
  Melting Point
  Density
  Bulk Density
  Autoignition temp
  Anal. Standard Form
  Potency
  Manufacturer designation
  Agency/Method
  Application(s)
  Solubility
  Physical Form
  Assay
Show More