• USA Home
  • Search Results

  • vanillinmeltingpointstandard1521512133511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Vanillin melting point standard

2 Products
Synonym:   4-Hydroxy-3-methoxybenzaldehyde, Vanillic aldehyde, Vanillin
Image of Vanillin melting point standard
CAS Number:
Linear Formula:
 4-(HO)C6H3-3-(OCH3)CHO
Molecular Weight:
 152.15
 Beilstein/REAXYS Number:
 472792
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
Show More