• USA Home
  • Search Results

  • vanillylalcohol1541649800011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Vanillyl alcohol

2 Products
Synonym:   4-Hydroxy-3-methoxybenzyl alcohol, NSC 3993, Vanillyl alcohol
Image of Vanillyl alcohol
CAS Number:
Linear Formula:
 HOC6H3(OCH3)CH2OH
Molecular Weight:
 154.16
 Beilstein/REAXYS Number:
 1910044
 Council of Europe no.:
 690
 EC Number:
 FEMA Number:
 3737
 Flavis number:
 2.213
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Assay
Show More