• USA Home
  • Search Results

  • vedaprofen282387110909611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Vedaprofen

3 Products
Synonym:   (±)-2-(4-Cyclohexylnaphthalen-1-yl)propionic acid
Image of Vedaprofen
CAS Number:
Empirical Formula (Hill Notation):
 C19H22O2
Molecular Weight:
 282.38
 Beilstein/REAXYS Number:
 5568831
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Anal. Standard Form
  Special Grade
  Application(s)
Show More