• USA Home
  • Search Results

  • venlafaxined6hydrochloridesolution31990106260612511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Venlafaxine-D6 hydrochloride solution

2 Products
Image of Venlafaxine-D<SUB>6</SUB> hydrochloride solution
CAS Number:
Empirical Formula (Hill Notation):
 C17D6H21NO2 · HCl
Molecular Weight:
 319.90
 EC Number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Special Grade
  Package quantity
  Manufacturer designation
  Feature
Show More