• USA Home
  • Search Results

  • verapamilhydrochloride4910615211411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(±)-Verapamil hydrochloride

5 Products
Synonym:   5-[N-(3,4-Dimethoxyphenylethyl)methylamino]-2-(3,4-dimethoxyphenyl)-2-isopropylvaleronitrile hydrochloride
Image of (±)-Verapamil hydrochloride
CAS Number:
Linear Formula:
 (CH3O)2C6H3CH2CH2N(CH3)(CH2)3C[C6H3(OCH3)2][CH(CH3)2]CN · HCl
Molecular Weight:
 491.06
 Beilstein/REAXYS Number:
 3647093
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
  Solubility
  Anal. Standard Form
  Physical Form
  Manufacturer designation
Show More