• USA Home
  • Search Results

  • vernolat20334192977711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Vernolat

1 Products
Synonym:   S-Propyl dipropylthiocarbamate
Image of Vernolat
CAS Number:
Empirical Formula (Hill Notation):
 C10H21NOS
Molecular Weight:
 203.34
 Beilstein/REAXYS Number:
 1767659
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More