• USA Home
  • Search Results

  • vigabatrin129166850686511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(±)-Vigabatrin

4 Products
Synonym:   (±)-γ-Vinyl-GABA, (±)-4-Aminohexenoic acid, (R,S)-4-Amino-5-hexenoic acid
Image of (±)-Vigabatrin
CAS Number:
Empirical Formula (Hill Notation):
 C6H11NO2
Molecular Weight:
 129.16
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More