• USA Home
  • Search Results

  • vincamine35444161790911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Vincamine

2 Products
Image of Vincamine
CAS Number:
Empirical Formula (Hill Notation):
 C21H26N2O3
Molecular Weight:
 354.44
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More