• USA Home
  • Search Results

  • vinylacetate860910805411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Vinyl acetate

5 Products
Synonym:   Acetic acid vinyl ester
Image of Vinyl acetate
CAS Number:
Empirical Formula (Hill Notation):
 C4H6O2
Molecular Weight:
 86.09
 EC Index Number
 607-023-00-0
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Package quantity
  Anal. Standard Form
  Physical Form
  Special Grade
  Agency/Method
Show More