• USA Home
  • Search Results

  • vitamink1450708480011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Vitamin K1

2 Products
Synonym:   2-Methyl-3-phytyl-1,4-naphthoquinone, 3-Phytylmenadione, Phylloquinone, Vitamin K1(20)
Image of Vitamin K<SUB>1</SUB>
CAS Number:
Empirical Formula (Hill Notation):
 C31H46O2
Molecular Weight:
 450.70
 Beilstein/REAXYS Number:
 2568816
 EC Number:
 MDL number:
 PubChem Substance ID:
 eCl@ss:
 34058015
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Color
  Physical Form
  Biological Source
Show More