• USA Home
  • Search Results

  • vitexin2orhamnoside578526482099111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Vitexin 2-O-rhamnoside

3 Products
Synonym:   2-O-Rhamnosylvitexin
Image of Vitexin 2-<I>O</I>-rhamnoside
CAS Number:
Empirical Formula (Hill Notation):
 C27H30O14
Molecular Weight:
 578.52
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Special Grade
Show More