• USA Home
  • Search Results

  • vitexin43238368193411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Vitexin

4 Products
Synonym:   4′,5,7-Trihydroxyflavone 8-C-glucoside, 8-C-Glucosylapigenin, Apigenin 8-C-glucoside, Orientoside
Image of Vitexin
CAS Number:
Empirical Formula (Hill Notation):
 C21H20O10
Molecular Weight:
 432.38
 Beilstein/REAXYS Number:
 67796
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Application(s)
  Assay
  Special Grade
Show More