• USA Home
  • Search Results

  • warfarin308338181211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Warfarin

4 Products
Synonym:   4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin, Coumafene
Image of Warfarin
CAS Number:
Empirical Formula (Hill Notation):
 C19H16O4
Molecular Weight:
 308.33
 Beilstein/REAXYS Number:
 8868198
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
  Manufacturer designation
  Application(s)
Show More