• USA Home
  • Search Results

  • water1802773218511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Water

30 Products
Synonym:   DNase-Free Water, Not DEPC Treated, Nuclease-Free Water, Protease-Free Water, RNase-Free Water
Image of Water
CAS Number:
Linear Formula:
 H2O
Molecular Weight:
 18.02
 Beilstein/REAXYS Number:
 2050024
 EC Number:
 MDL number:
 NACRES:
 NA.31
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  UV-Absorption Condition
  Product Line
  Quality clarifier
  Ice Shipping
  Shelf Life
  Conductivity
  UV-Absorption Values
  Sterilization
  Absorption
  Vapor density vs air
  Special Grade
  pH Value
  Application(s)
  Vapor pressure
Show More

Product #
Description
W4502 Water, Nuclease-Free Water, for Molecular Biology,
W3500 Water, sterile-filtered, BioReagent, suitable for cell culture,
270733 Water, for HPLC,
34877 Water, HPLC Plus,
1.15333 Water, for chromatography (LC-MS Grade) LiChrosolv®,
95284 Water, for molecular biology, sterile filtered,
00612 Water, for ion chromatography,
38796 Water, deionized,
W1503 Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture,
900682 Water, for UHPLC, for mass spectrometry,
WX0001 Water, OmniSolv® LC-MS,
78533 Water, for TOC analysis,
1.03728 Water, for UHPLC-MS LiChrosolv®,
14211 Water, ACS reagent, for ultratrace analysis,
W3513 Water, BioPerformance Certified,
W1754 Water, PCR Reagent,
95289 Water, for cell biology, sterile ultrafiltered,
WX0004 Water, gradient grade, Meets Reagent Specifications for testing USP/NF monographs OmniSolv®,
1.01262 Water, Ultrapur,
1.16754 Water, for analysis EMSURE®,
1.00473 Water, for inorganic trace analysis Suprapur®,
WX0008 Water, HPLC,
95280 Water, tested according to Ph. Eur.,
320072 Water, ACS reagent,
WX0003 Water, OmniTrace Ultra,
95283 Water, for HPCE, for luminescence, for UV-spectroscopy,
1.00577 Water, for headspace gas chromatography SupraSolv®,
1.03702 Water, for gas chromatography MS SupraSolv®,
1.02699 Water, for gas chromatography SupraSolv®,
99053 Water, deionized,