• USA Home
  • Search Results

  • water18o20021431442211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Water-18O

7 Products
Synonym:   18O Water
Image of Water-<SUP>18</SUP>O
CAS Number:
Linear Formula:
 H218O
Molecular Weight:
 20.02
 MDL number:
 NACRES:
 NA.12
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Isotopic Purity
  Compatibility
  Molecular weight
Show More