• USA Home
  • Search Results

  • wintergreenoil123459004528611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Wintergreen oil

1 Products
Image of Wintergreen oil
CAS Number:
 EC Number:
 FEMA Number:
 3113
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More