• USA Home
  • Search Results

  • xanthine152116989611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Xanthine

3 Products
Synonym:   2,6-Dihydroxypurine
Image of Xanthine
CAS Number:
Empirical Formula (Hill Notation):
 C5H4N4O2
Molecular Weight:
 152.11
 Beilstein/REAXYS Number:
 8733
 EC Number:
 MDL number:
 NACRES:
 NA.51
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Physical Form
  Biological Source
  Assay
Show More