• USA Home
  • Search Results

  • xanthohumol35440675458111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Xanthohumol

2 Products
Synonym:   (E)-1-[2,4-Dihydroxy-6-methoxy-3-(3-methyl-2-butenyl)phenyl]-3-(4-hydroxyphenyl)propenone, 2′,4,4′-Trihydroxy-6′-methoxy-3′-prenylchalcone
Image of Xanthohumol
CAS Number:
Empirical Formula (Hill Notation):
 C21H22O5
Molecular Weight:
 354.40
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Application(s)
Show More