• USA Home
  • Search Results

  • xylenecyanolff53861265017111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Xylene Cyanol FF

2 Products
Synonym:   Acid blue 147, Cyanol FF, XC
Image of Xylene Cyanol FF
CAS Number:
Empirical Formula (Hill Notation):
 C25H27N2NaO6S2
Molecular Weight:
 538.61
 Colour Index Number:
 42135
 EC Number:
 MDL number:
 NACRES:
 NA.52
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Physical Form
  Application(s)
Show More