• USA Home
  • Search Results

  • xylenolorangetetrasodiumsalt76058361843711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Xylenol orange tetrasodium salt

2 Products
Synonym:   3,3′′-Bis[bis(carboxymethyl)aminoethyl] cresol sulfone phtrane sodium salt, Xylenol orange tetrasodium salt
Image of Xylenol orange tetrasodium salt
CAS Number:
Empirical Formula (Hill Notation):
 C31H28N2Na4O13S
Molecular Weight:
 760.58
 EC Index Number:
 222-805-8
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Special Grade
Show More