• USA Home
  • Search Results

  • xylitol152158799011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Xylitol

6 Products
Synonym:   Xylite
Image of Xylitol
CAS Number:
Linear Formula:
 HOCH2[CH(OH)]3CH2OH
Molecular Weight:
 152.15
 Beilstein/REAXYS Number:
 1720523
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Manufacturer designation
  Assay
  Anal. Standard Form
  Application(s)
  Special Grade
Show More