• USA Home
  • Search Results

  • yeastextract12345801301211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Yeast extract

7 Products
Synonym:   Extract of yeast, Yeast extract
Image of Yeast extract
CAS Number:
 EC Index Number:
 232-387-9
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Media Suitability
  Chemical Composition
  Density
  Bulk Density
  Autoignition temp
  Shelf Life
  Special Grade
  Loss
  Physical Form
  pH Value
  Application(s)
  Solubility
  Assay
Show More