• USA Home
  • Search Results

  • ylangylangoil12345800681311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ylang-ylang oil

5 Products
CAS Number:
 FEMA Number:
 3119
 MDL number:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Allergen
  Biological Source
  Refractive Index
  Document Links
  Organoleptic property
  Density
  Special Grade
Show More