• USA Home
  • Search Results

  • yttriumiiioxidedispersion22581131436911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Yttrium(III) oxide, dispersion

1 Products
Synonym:   Yttria
Image of Yttrium(III) oxide, dispersion
CAS Number:
Linear Formula:
 Y2O3
Molecular Weight:
 225.81
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Particle Dimension
  Concentration
  Assay
  Physical Form
Show More