• USA Home
  • Search Results

  • zdehydroalaome235242114917711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Z-Dehydro-Ala-OMe

1 Products
Synonym:   N-Cbz-Dehydroalanine methyl ester, Z-Dehydroalanine methyl ester
Image of Z-Dehydro-Ala-<I>O</I>Me
CAS Number:
Linear Formula:
 C6H5CH2O2CNHC(=CH2)CO2CH3
Molecular Weight:
 235.24
 Beilstein/REAXYS Number:
 2056075
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More