• USA Home
  • Search Results

  • zglnoh28028265064811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Z-Gln-OH

1 Products
Synonym:   N-Cbz-L-glutamine, Carbobenzyloxy-L-glutamine, Z-L-Glutamine
Image of Z-Gln-OH
CAS Number:
Linear Formula:
 NH2COCH2CH2CH(NHCOOCH2C6H5)COOH
Molecular Weight:
 280.28
 Beilstein/REAXYS Number:
 2061271
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
162647 Z-Gln-OH, 99%,