• USA Home
  • Search Results

  • zincchloride13630764685711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Zinc chloride

14 Products
Synonym:   Dichlorozinc
Image of Zinc chloride
CAS Number:
Linear Formula:
 ZnCl2
Molecular Weight:
 136.30
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Application(s)
  Particle Dimension
  Melting Point
  Quality clarifier
  Purity Grade
  Potency
  Density
  Bulk Density
  Vapor pressure
  Product Line
  Reaction Suitability
  Agency/Method
  Solubility
  pH Value
  Physical Form
  Special Grade
Show More

Product #
Description
208086 Zinc chloride, reagent grade, ≥98%,
793523 Zinc chloride, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%,
229997 Zinc chloride, 99.999% trace metals basis,
Z0152 Zinc chloride, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture,
429430 Zinc chloride, anhydrous, powder, ≥99.995% trace metals basis,
746355 Zinc chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥97%,
31650 Zinc chloride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥98%,
211273 Zinc chloride, ACS reagent, ≥97%,
1.08816 Zinc chloride, for analysis EMSURE® ACS,ISO,Reag. Ph Eur,
14422 Zinc chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98-100.5%,
450111 Zinc chloride, AnhydroBeads, amorphous, −10 mesh, 99.99% trace metals basis,
456845 Zinc chloride, AnhydroBeads, amorphous, −10 mesh, 99.999% trace metals basis,
1.08815 Zinc chloride, EMPROVE® API Ph Eur,BP,USP,
1.16763 Zinc chloride, Technipur,