• USA Home
  • Search Results

  • zincchloridesolution13630764685711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Zinc chloride solution

3 Products
Synonym:   Dichlorozinc
Image of Zinc chloride solution
CAS Number:
Linear Formula:
 ZnCl2
Molecular Weight:
 136.30
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Density
Show More