• USA Home
  • Search Results

  • zincnitratehexahydrate297491019618611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Zinc nitrate hexahydrate

2 Products
Image of Zinc nitrate hexahydrate
CAS Number:
Linear Formula:
 Zn(NO3)2 · 6H2O
Molecular Weight:
 297.49
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Solubility
  Purity Grade
Show More