• USA Home
  • Search Results

  • zincoxide8139131413211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Zinc oxide

15 Products
Image of Zinc oxide
CAS Number:
Linear Formula:
 ZnO
Molecular Weight:
 81.39
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Restriction
  pH Value
  Surface Dimension
  Purity Grade
  Potency
  Melting Point
  Density
  Bulk Density
  Product Line
  Particle Dimension
  Composite Dimension
  Reaction Suitability
  Application(s)
  Agency/Method
  Special Grade
  Assay
Show More

Product #
Description
544906 Zinc oxide, nanopowder, <100 nm particle size,
96479 Zinc oxide, puriss. p.a., ACS reagent, ≥99.0% (KT),
677450 Zinc oxide, nanopowder, <50 nm particle size (BET), >97%,
205532 Zinc oxide, ReagentPlus®, powder, <5 μm particle size, 99.9%,
255750 Zinc oxide, 99.99% trace metals basis,
204951 Zinc oxide, 99.999% trace metals basis,
1724747 Zinc oxide, United States Pharmacopeia (USP) Reference Standard,
14439 Zinc oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. for dried substance),
773980 Zinc oxide, nanowires, size × L × 300 nm,
93632 Zinc oxide, analytical standard,
773999 Zinc oxide, nanowires, size × L × 1 μm,
1.08849 Zinc oxide, for analysis EMSURE® ACS,Reag. Ph Eur,
774006 Zinc oxide, nanowires, size × L × 4-5 μm,
PHR2724 Zinc oxide, Pharmaceutical Secondary Standard; Certified Reference Material, certified reference material, pharmaceutical secondary standard, pkg of 500 mg, New
1.08846 Zinc oxide, EMPROVE® ESSENTIAL Ph Eur,BP,JP,USP,