• USA Home
  • Search Results

  • zineb275751212267711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Zineb

1 Products
Synonym:   Zinc-ethylenebis(dithiocarbamate)
Image of Zineb
CAS Number:
Empirical Formula (Hill Notation):
 C4H6N2S4Zn
Molecular Weight:
 275.75
 Beilstein/REAXYS Number:
 4165797
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More