• USA Home
  • Search Results

  • zingerone1942312248511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Zingerone

1 Products
Synonym:   Vanillylacetone, Zingerone
Image of Zingerone
CAS Number:
Linear Formula:
 4-(HO)C6H3-3-(OCH3)CH2CH2COCH3
Molecular Weight:
 194.23
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More