• USA Home
  • Search Results

  • ziram3058213730411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ziram

1 Products
Synonym:   Dimethyldithiocarbamic acid zinc salt, Zinc dimethyldithiocarbamate, Ziram
Image of Ziram
CAS Number:
Linear Formula:
 (CH3)2NCSSZnSCSN(CH3)2
Molecular Weight:
 305.82
 Beilstein/REAXYS Number:
 3707008
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More